Product Details

SNP ID
rs141108014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96981603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAACACCTTTGCGCCAGGATTT[A/G]GAGGCAAATGAACAAGATGCAAGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf12 PubMed Links

Gene Details

Gene
C10orf12
Gene Name
chromosome 10 open reading frame 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015652.2 320 Silent Mutation TTA,TTG L71L NP_056467.2

View Full Product Details