Product Details

SNP ID
rs141161147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101227296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCGTCCACGTCGATCTCTTCGT[C/T]TTCCTCGTCCTCCGAGCAGTCCTGG
Phenotype
MIM: 604255
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LBX1 PubMed Links

Gene Details

Gene
LBX1
Gene Name
ladybird homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006562.4 965 Missense Mutation AAC,GAC N274D NP_006553.2
Gene
LBX1-AS1
Gene Name
LBX1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details