Product Details

SNP ID
rs141171117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5953184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCCTCCATGGCTTCCATTTCAA[C/T]GCTGGCCAGCGGGGGAGTTTGCCTG
Phenotype
MIM: 601070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL15RA PubMed Links

Gene Details

Gene
IL15RA
Gene Name
interleukin 15 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243539.1 1228 Missense Mutation NP_001230468.1
NM_001256765.1 1228 Missense Mutation NP_001243694.1
NM_002189.3 1228 Missense Mutation NP_002180.1
NM_172200.2 1228 Missense Mutation NP_751950.2
XM_011519461.2 1228 Intron XP_011517763.1
XM_011519462.2 1228 Intron XP_011517764.1
XM_011519463.2 1228 Intron XP_011517765.1
XM_011519464.2 1228 Intron XP_011517766.1
XM_011519465.1 1228 Missense Mutation XP_011517767.1
XM_011519466.2 1228 UTR 3 XP_011517768.1
XM_011519467.2 1228 Intron XP_011517769.1
XM_011519468.1 1228 Intron XP_011517770.1
XM_011519469.1 1228 Missense Mutation XP_011517771.1
XM_011519470.2 1228 Intron XP_011517772.1
XM_011519471.2 1228 Intron XP_011517773.1
XM_011519472.2 1228 Intron XP_011517774.1
XM_011519474.2 1228 Intron XP_011517776.1
XM_011519475.2 1228 Intron XP_011517777.1
XM_011519476.2 1228 Missense Mutation XP_011517778.1
XM_011519477.2 1228 UTR 3 XP_011517779.1
XM_017016195.1 1228 Missense Mutation XP_016871684.1
XM_017016196.1 1228 Intron XP_016871685.1
XM_017016197.1 1228 Intron XP_016871686.1
XM_017016198.1 1228 Missense Mutation XP_016871687.1
XM_017016199.1 1228 Intron XP_016871688.1
XM_017016200.1 1228 Intron XP_016871689.1
XM_017016201.1 1228 UTR 3 XP_016871690.1
XM_017016202.1 1228 UTR 3 XP_016871691.1
XM_017016203.1 1228 Missense Mutation XP_016871692.1

View Full Product Details