Product Details

SNP ID
rs141402057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:59794550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAAGACATTCCAGCTCTAGTCA[C/G]TGAAGTTGGTGGCGTGAAACCAACC
Phenotype
MIM: 601985
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC6 PubMed Links

Gene Details

Gene
CCDC6
Gene Name
coiled-coil domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005436.4 1385 Missense Mutation CTG,GTG L385V NP_005427.2

View Full Product Details