Product Details

SNP ID
rs141480317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101002824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCACCTACATCAATGAGGACTTC[C/T]GGACAGAGTCACCCCCCAGCCCAAG
Phenotype
MIM: 606075 MIM: 610454 MIM: 612971
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf2 PubMed Links

Gene Details

Gene
C10orf2
Gene Name
chromosome 10 open reading frame 2
There are no transcripts associated with this gene.

Gene
LZTS2
Gene Name
leucine zipper tumor suppressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318099.1 498 Missense Mutation CGG,TGG R96W NP_001305028.1
NM_001318100.1 498 Missense Mutation CGG,TGG R96W NP_001305029.1
NM_001318101.1 498 Missense Mutation CGG,TGG R96W NP_001305030.1
NM_032429.3 498 Missense Mutation CGG,TGG R96W NP_115805.1
XM_005270222.4 498 Missense Mutation CGG,TGG R96W XP_005270279.1
XM_017016780.1 498 Missense Mutation CGG,TGG R96W XP_016872269.1
XM_017016781.1 498 Missense Mutation CGG,TGG R96W XP_016872270.1
Gene
PDZD7
Gene Name
PDZ domain containing 7
There are no transcripts associated with this gene.

View Full Product Details