Product Details

SNP ID
rs141559236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:132808472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAGTCTTCTGGTCTAAGCAACT[C/G]AGCACCAGCGCCGCCAAGGGGAGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFAP46 PubMed Links

Gene Details

Gene
CFAP46
Gene Name
cilia and flagella associated protein 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001200049.2 8198 Silent Mutation CTC,CTG L2699L NP_001186978.2

View Full Product Details