Product Details

SNP ID
rs141631018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102676934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGACATTTTTGCAGACCCAGTTC[A/G]TGCCATCCTTTGAGGGAAATGGGCA
Phenotype
MIM: 604695
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL3 PubMed Links

Gene Details

Gene
ARL3
Gene Name
ADP ribosylation factor like GTPase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004311.3 853 Missense Mutation ACG,ATG T170M NP_004302.1
XM_017016260.1 853 Missense Mutation ACG,ATG T170M XP_016871749.1

View Full Product Details