Product Details

SNP ID
rs141964073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110207935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCGGCCCTGCCCGAGGACCCC[A/G]CTGGGGCCAAGCCCAGGTGCCCCTT
Phenotype
MIM: 600020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MXI1 PubMed Links

Gene Details

Gene
MXI1
Gene Name
MAX interactor 1, dimerization protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008541.1 331 Intron NP_001008541.1
NM_005962.4 331 Intron NP_005953.4
NM_130439.3 331 Missense Mutation ACT,GCT T43A NP_569157.2

View Full Product Details