Product Details

SNP ID
rs142052016
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:42796744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACAGTGAAATGGATTTGCCAGC[A/G]TTTGCTGACAGTGACGATGACCTTG
Phenotype
MIM: 611448
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BMS1 PubMed Links

Gene Details

Gene
BMS1
Gene Name
BMS1, ribosome biogenesis factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014753.3 1589 Silent Mutation GCA,GCG A500A NP_055568.3
XM_005271846.3 1589 Silent Mutation GCA,GCG A500A XP_005271903.1
XM_005271848.3 1589 Silent Mutation GCA,GCG A500A XP_005271905.1
XM_005271849.3 1589 Silent Mutation GCA,GCG A500A XP_005271906.1
XM_011540402.2 1589 Silent Mutation GCA,GCG A500A XP_011538704.1
XM_011540403.2 1589 Silent Mutation GCA,GCG A89A XP_011538705.1

View Full Product Details