Product Details

SNP ID
rs142120325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:16825012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTAATCGGAATGCGGATGGACGC[C/T]GTGCATAATCAGCATGAAATTTTAT
Phenotype
MIM: 602997 MIM: 179555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CUBN PubMed Links

Gene Details

Gene
CUBN
Gene Name
cubilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081.3 7250 Missense Mutation CAG,CGG Q3612R NP_001072.2
XM_011519708.2 7250 Intron XP_011518010.1
XM_011519709.2 7250 Missense Mutation CAG,CGG Q2274R XP_011518011.1
XM_011519710.2 7250 Missense Mutation CAG,CGG Q2266R XP_011518012.1
XM_011519711.2 7250 Missense Mutation CAG,CGG Q2226R XP_011518013.1
Gene
RSU1
Gene Name
Ras suppressor protein 1
There are no transcripts associated with this gene.

View Full Product Details