Product Details

SNP ID
rs142205964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100042551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCACAGGGCTTACTTGAGGGATG[C/T]TTCTTTTGAGGTGGAAGTTAACCTG
Phenotype
MIM: 603103
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPN1 PubMed Links

Gene Details

Gene
CPN1
Gene Name
carboxypeptidase N subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308.2 1513 Missense Mutation AAC,AGC N418S NP_001299.1

View Full Product Details