Product Details

SNP ID
rs142276906
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102918840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAGACGTGGTCCCGCATCGCCTT[C/T]ACCGAGCACGAGCGGCGGCGCCACA
Phenotype
MIM: 607803
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CNNM2 PubMed Links

Gene Details

Gene
CNNM2
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017649.4 548 Silent Mutation TTC,TTT F120F NP_060119.3
NM_199076.2 548 Silent Mutation TTC,TTT F120F NP_951058.1
NM_199077.2 548 Silent Mutation TTC,TTT F120F NP_951059.1
XM_005269933.4 548 Silent Mutation TTC,TTT F120F XP_005269990.1
XM_006717908.2 548 Silent Mutation TTC,TTT F120F XP_006717971.1
XM_011539911.2 548 Silent Mutation TTC,TTT F120F XP_011538213.1
Gene
LOC107984265
Gene Name
translation initiation factor IF-2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017017027.1 548 Intron XP_016872516.1

View Full Product Details