Product Details

SNP ID
rs142455548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89417382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATAACCTATTGGCCTATGTGAA[A/C]CACCTAAAAGGCCAAAATAAAGACG
Phenotype
MIM: 616135
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IFIT5 PubMed Links

Gene Details

Gene
IFIT5
Gene Name
interferon induced protein with tetratricopeptide repeats 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012420.2 414 Silent Mutation AAA,AAC K61N NP_036552.1

View Full Product Details