Product Details

SNP ID
rs142581169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101579667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAAGTGTGCAGAGCCGGTGAAG[G/T]AGAGCAGGGCACAGGCAAACTCGCT
Phenotype
MIM: 616467 MIM: 606343
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DPCD PubMed Links

Gene Details

Gene
DPCD
Gene Name
deleted in primary ciliary dyskinesia homolog (mouse)
There are no transcripts associated with this gene.

Gene
POLL
Gene Name
polymerase (DNA) lambda
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174084.1 2009 Missense Mutation NP_001167555.1
NM_001174085.1 2009 Missense Mutation NP_001167556.1
NM_001308382.1 2009 Missense Mutation NP_001295311.1
NM_013274.3 2009 Missense Mutation NP_037406.1
XM_006717775.1 2009 Missense Mutation XP_006717838.1
XM_006717776.1 2009 Missense Mutation XP_006717839.1
XM_006717777.1 2009 Missense Mutation XP_006717840.1
XM_011539650.1 2009 Missense Mutation XP_011537952.1
XM_011539651.1 2009 Missense Mutation XP_011537953.1
XM_011539652.1 2009 Missense Mutation XP_011537954.1
XM_011539653.1 2009 Missense Mutation XP_011537955.1
XM_011539654.1 2009 Missense Mutation XP_011537956.1
XM_011539655.1 2009 Missense Mutation XP_011537957.1
XM_011539656.1 2009 Missense Mutation XP_011537958.1
XM_011539657.1 2009 Missense Mutation XP_011537959.1
XM_011539659.1 2009 Missense Mutation XP_011537961.1
XM_011539660.1 2009 Missense Mutation XP_011537962.1
XM_011539662.1 2009 Missense Mutation XP_011537964.1
XM_011539663.1 2009 Missense Mutation XP_011537965.1
XM_011539664.1 2009 Missense Mutation XP_011537966.1
XM_011539665.2 2009 Missense Mutation XP_011537967.1
XM_011539666.1 2009 Missense Mutation XP_011537968.1
XM_011539667.1 2009 Missense Mutation XP_011537969.1
XM_017016084.1 2009 Missense Mutation XP_016871573.1
XM_017016085.1 2009 Missense Mutation XP_016871574.1
XM_017016086.1 2009 Missense Mutation XP_016871575.1
XM_017016087.1 2009 Missense Mutation XP_016871576.1
XM_017016088.1 2009 Intron XP_016871577.1
XM_017016089.1 2009 Missense Mutation XP_016871578.1
XM_017016090.1 2009 UTR 3 XP_016871579.1
XM_017016091.1 2009 Missense Mutation XP_016871580.1
XM_017016092.1 2009 Intron XP_016871581.1
XM_017016093.1 2009 Missense Mutation XP_016871582.1

View Full Product Details