Product Details
- SNP ID
-
rs142956872
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:80157706 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTCTGATCTGATGTCCAGGAGGT[C/T]GGTCTCGCTGCGAGACACCATGATG
- Phenotype
-
MIM: 602572
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA11
PubMed Links
Gene Details
- Gene
- ANXA11
- Gene Name
- annexin A11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001157.2 |
1949 |
Missense Mutation |
AAC,GAC |
N465D |
NP_001148.1 |
NM_001278407.1 |
1949 |
Missense Mutation |
AAC,GAC |
N465D |
NP_001265336.1 |
NM_001278408.1 |
1949 |
Missense Mutation |
AAC,GAC |
N465D |
NP_001265337.1 |
NM_001278409.1 |
1949 |
Missense Mutation |
AAC,GAC |
N432D |
NP_001265338.1 |
NM_145868.1 |
1949 |
Missense Mutation |
AAC,GAC |
N465D |
NP_665875.1 |
NM_145869.1 |
1949 |
Missense Mutation |
AAC,GAC |
N465D |
NP_665876.1 |
XM_005269741.4 |
1949 |
Missense Mutation |
AAC,GAC |
N565D |
XP_005269798.1 |
XM_005269742.1 |
1949 |
Missense Mutation |
AAC,GAC |
N465D |
XP_005269799.1 |
XM_006717813.1 |
1949 |
Missense Mutation |
AAC,GAC |
N465D |
XP_006717876.1 |
XM_006717814.3 |
1949 |
Missense Mutation |
AAC,GAC |
N465D |
XP_006717877.1 |
XM_011539735.1 |
1949 |
Missense Mutation |
AAC,GAC |
N465D |
XP_011538037.1 |
XM_011539736.2 |
1949 |
Intron |
|
|
XP_011538038.1 |
View Full Product Details