Product Details

SNP ID
rs143097855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27504394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGACGAGGACGTGCTAACCACC[C/T]TGAAGATCCTCATCATCGGCGAGAG
Phenotype
MIM: 602207
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB18 PubMed Links

Gene Details

Gene
RAB18
Gene Name
RAB18, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256410.1 221 Silent Mutation CTG,TTG L9L NP_001243339.1
NM_001256411.1 221 Silent Mutation CTG,TTG L9L NP_001243340.1
NM_001256412.1 221 Silent Mutation CTG,TTG L9L NP_001243341.1
NM_001256415.1 221 Silent Mutation CTG,TTG L9L NP_001243344.1
NM_021252.4 221 Silent Mutation CTG,TTG L9L NP_067075.1

View Full Product Details