Product Details

SNP ID
rs143121841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100048762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAGCTCAGCCTAACTCACCAAC[A/G]TTGGTTCCGCAGGACCCACGGTCAC
Phenotype
MIM: 603103
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPN1 PubMed Links

Gene Details

Gene
CPN1
Gene Name
carboxypeptidase N subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308.2 1486 Missense Mutation ACG,ATG T409M NP_001299.1

View Full Product Details