Product Details

SNP ID
rs143455921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99877193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGATATAATTGGAGGGAACGTA[A/G]CCCTTCTTCCCGTTAACCTCAGCTA
Phenotype
MIM: 611282
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNMBP PubMed Links

Gene Details

Gene
DNMBP
Gene Name
dynamin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318326.1 4684 Silent Mutation GGC,GGT G1196G NP_001305255.1
NM_001318327.1 4684 Silent Mutation GGC,GGT G852G NP_001305256.1
NM_015221.3 4684 Silent Mutation GGC,GGT G1564G NP_056036.1
XM_006717735.2 4684 Silent Mutation GGC,GGT G1521G XP_006717798.1
XM_011539559.2 4684 Silent Mutation GGC,GGT G1564G XP_011537861.1

View Full Product Details