Product Details
- SNP ID
-
rs143966324
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:5953152 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCTCTGCTGCTGGTCCCCCAAGT[C/T]ACCGGCAGAGCCTCCATGGCTTCCA
- Phenotype
-
MIM: 601070
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL15RA
PubMed Links
Gene Details
- Gene
- IL15RA
- Gene Name
- interleukin 15 receptor subunit alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001243539.1 |
1260 |
Silent Mutation |
GTA,GTG |
V213V |
NP_001230468.1 |
NM_001256765.1 |
1260 |
Silent Mutation |
GTA,GTG |
V335V |
NP_001243694.1 |
NM_002189.3 |
1260 |
Silent Mutation |
GTA,GTG |
V249V |
NP_002180.1 |
NM_172200.2 |
1260 |
Silent Mutation |
GTA,GTG |
V216V |
NP_751950.2 |
XM_011519461.2 |
1260 |
Intron |
|
|
XP_011517763.1 |
XM_011519462.2 |
1260 |
Intron |
|
|
XP_011517764.1 |
XM_011519463.2 |
1260 |
Intron |
|
|
XP_011517765.1 |
XM_011519464.2 |
1260 |
Intron |
|
|
XP_011517766.1 |
XM_011519465.1 |
1260 |
Silent Mutation |
GTA,GTG |
V358V |
XP_011517767.1 |
XM_011519466.2 |
1260 |
UTR 3 |
|
|
XP_011517768.1 |
XM_011519467.2 |
1260 |
Intron |
|
|
XP_011517769.1 |
XM_011519468.1 |
1260 |
Intron |
|
|
XP_011517770.1 |
XM_011519469.1 |
1260 |
Silent Mutation |
GTA,GTG |
V325V |
XP_011517771.1 |
XM_011519470.2 |
1260 |
Intron |
|
|
XP_011517772.1 |
XM_011519471.2 |
1260 |
Intron |
|
|
XP_011517773.1 |
XM_011519472.2 |
1260 |
Intron |
|
|
XP_011517774.1 |
XM_011519474.2 |
1260 |
Intron |
|
|
XP_011517776.1 |
XM_011519475.2 |
1260 |
Intron |
|
|
XP_011517777.1 |
XM_011519476.2 |
1260 |
Silent Mutation |
GTA,GTG |
V186V |
XP_011517778.1 |
XM_011519477.2 |
1260 |
UTR 3 |
|
|
XP_011517779.1 |
XM_017016195.1 |
1260 |
Silent Mutation |
GTA,GTG |
V307V |
XP_016871684.1 |
XM_017016196.1 |
1260 |
Intron |
|
|
XP_016871685.1 |
XM_017016197.1 |
1260 |
Intron |
|
|
XP_016871686.1 |
XM_017016198.1 |
1260 |
Silent Mutation |
GTA,GTG |
V242V |
XP_016871687.1 |
XM_017016199.1 |
1260 |
Intron |
|
|
XP_016871688.1 |
XM_017016200.1 |
1260 |
Intron |
|
|
XP_016871689.1 |
XM_017016201.1 |
1260 |
UTR 3 |
|
|
XP_016871690.1 |
XM_017016202.1 |
1260 |
UTR 3 |
|
|
XP_016871691.1 |
XM_017016203.1 |
1260 |
Silent Mutation |
GTA,GTG |
V180V |
XP_016871692.1 |
View Full Product Details