Product Details

SNP ID
rs143979325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:22316426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACGGCGCGCTCACAATGGAGCT[C/G]TCGGAGTCTGTGCAGAAAGGCTTCC
Phenotype
MIM: 164831 MIM: 616700
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BMI1 PubMed Links

Gene Details

Gene
BMI1
Gene Name
BMI1 proto-oncogene, polycomb ring finger
There are no transcripts associated with this gene.

Gene
COMMD3
Gene Name
COMM domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012071.3 44 Silent Mutation CTC,CTG L3L NP_036203.1
Gene
COMMD3-BMI1
Gene Name
COMMD3-BMI1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204062.1 44 Silent Mutation CTC,CTG L3L NP_001190991.1

View Full Product Details