Product Details

SNP ID
rs144127339
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:114432217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAATTGTAAAAATCAAATAAAA[C/T]TGAGGCTGGCATAGGAAGTACAGAT
Phenotype
MIM: 602330
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ABLIM1 PubMed Links

Gene Details

Gene
ABLIM1
Gene Name
actin binding LIM protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003407.1 7139 UTR 3 NP_001003407.1
NM_001003408.1 7139 UTR 3 NP_001003408.1
NM_001322882.1 7139 UTR 3 NP_001309811.1
NM_001322883.1 7139 UTR 3 NP_001309812.1
NM_001322884.1 7139 UTR 3 NP_001309813.1
NM_001322885.1 7139 UTR 3 NP_001309814.1
NM_001322886.1 7139 UTR 3 NP_001309815.1
NM_001322887.1 7139 UTR 3 NP_001309816.1
NM_001322888.1 7139 UTR 3 NP_001309817.1
NM_001322889.1 7139 UTR 3 NP_001309818.1
NM_001322890.1 7139 UTR 3 NP_001309819.1
NM_001322891.1 7139 UTR 3 NP_001309820.1
NM_001322892.1 7139 UTR 3 NP_001309821.1
NM_001322893.1 7139 UTR 3 NP_001309822.1
NM_001322894.1 7139 UTR 3 NP_001309823.1
NM_001322895.1 7139 UTR 3 NP_001309824.1
NM_001322896.1 7139 UTR 3 NP_001309825.1
NM_001322897.1 7139 UTR 3 NP_001309826.1
NM_001322898.1 7139 UTR 3 NP_001309827.1
NM_001322899.1 7139 UTR 3 NP_001309828.1
NM_001322900.1 7139 UTR 3 NP_001309829.1
NM_002313.5 7139 UTR 3 NP_002304.3
NM_006720.3 7139 UTR 3 NP_006711.3
XM_005269824.2 7139 UTR 3 XP_005269881.1
XM_005269826.1 7139 UTR 3 XP_005269883.1
XM_005269827.4 7139 UTR 3 XP_005269884.1
XM_006717837.1 7139 UTR 3 XP_006717900.1
XM_006717846.3 7139 UTR 3 XP_006717909.1
XM_006717847.1 7139 UTR 3 XP_006717910.1
XM_011539801.1 7139 UTR 3 XP_011538103.1
XM_011539802.1 7139 UTR 3 XP_011538104.1
XM_011539803.1 7139 UTR 3 XP_011538105.1
XM_011539805.1 7139 UTR 3 XP_011538107.1
XM_017016242.1 7139 UTR 3 XP_016871731.1
XM_017016243.1 7139 UTR 3 XP_016871732.1
XM_017016244.1 7139 UTR 3 XP_016871733.1
XM_017016245.1 7139 UTR 3 XP_016871734.1
XM_017016246.1 7139 UTR 3 XP_016871735.1
XM_017016247.1 7139 UTR 3 XP_016871736.1
XM_017016248.1 7139 UTR 3 XP_016871737.1
XM_017016249.1 7139 UTR 3 XP_016871738.1
XM_017016250.1 7139 UTR 3 XP_016871739.1
XM_017016251.1 7139 UTR 3 XP_016871740.1
XM_017016252.1 7139 UTR 3 XP_016871741.1
XM_017016253.1 7139 UTR 3 XP_016871742.1
XM_017016254.1 7139 UTR 3 XP_016871743.1
XM_017016255.1 7139 UTR 3 XP_016871744.1
XM_017016256.1 7139 UTR 3 XP_016871745.1
XM_017016257.1 7139 Intron XP_016871746.1
XM_017016258.1 7139 UTR 3 XP_016871747.1
XM_017016259.1 7139 UTR 3 XP_016871748.1

View Full Product Details