Product Details
- SNP ID
-
rs144148304
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:77792493 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTTATTTTGTTCTTGATTGACCA[C/T]TGCCAAGATCTGAGTGCAAATGCTT
- Phenotype
-
MIM: 604090
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DLG5
PubMed Links
Gene Details
- Gene
- DLG5
- Gene Name
- discs large MAGUK scaffold protein 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004747.3 |
4720 |
Missense Mutation |
ATG,GTG |
M1903V |
NP_004738.3 |
XM_005270276.4 |
4720 |
Missense Mutation |
ATG,GTG |
M1899V |
XP_005270333.1 |
XM_006718056.3 |
4720 |
Missense Mutation |
ATG,GTG |
M1563V |
XP_006718119.1 |
XM_006718057.3 |
4720 |
Intron |
|
|
XP_006718120.1 |
XM_011540341.2 |
4720 |
Missense Mutation |
ATG,GTG |
M1844V |
XP_011538643.1 |
XM_011540342.1 |
4720 |
Missense Mutation |
ATG,GTG |
M1813V |
XP_011538644.1 |
XM_011540344.2 |
4720 |
Missense Mutation |
ATG,GTG |
M1791V |
XP_011538646.1 |
XM_011540345.1 |
4720 |
Missense Mutation |
ATG,GTG |
M1748V |
XP_011538647.1 |
XM_011540346.2 |
4720 |
Intron |
|
|
XP_011538648.1 |
XM_011540347.2 |
4720 |
Missense Mutation |
ATG,GTG |
M1597V |
XP_011538649.1 |
XM_017016913.1 |
4720 |
Missense Mutation |
ATG,GTG |
M1809V |
XP_016872402.1 |
XM_017016914.1 |
4720 |
Missense Mutation |
ATG,GTG |
M1793V |
XP_016872403.1 |
XM_017016915.1 |
4720 |
Missense Mutation |
ATG,GTG |
M1597V |
XP_016872404.1 |
View Full Product Details