Product Details

SNP ID
rs144307831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97767758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCACCAGCCGCTTGGTGTCCTTG[C/T]GCTTCAGGGGGCCCGGCTTGAGCAG
Phenotype
MIM: 604158 MIM: 610243
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SFRP5 PubMed Links

Gene Details

Gene
SFRP5
Gene Name
secreted frizzled related protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003015.3 876 Missense Mutation CAC,CGC H237R NP_003006.2
Gene
ZFYVE27
Gene Name
zinc finger FYVE-type containing 27
There are no transcripts associated with this gene.

View Full Product Details