Product Details
- SNP ID
-
hCV160591618
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:47300935 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCGGCAGGGCGCGCGGCCGGGAG[G/A]GGGCAACACGGTCCGCAGCTTCAGG
- Phenotype
-
MIM: 601361
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GDF10
PubMed Links
Gene Details
- Gene
- GDF10
- Gene Name
- growth differentiation factor 10
There are no transcripts associated with this gene.
View Full Product Details