Product Details

SNP ID
rs144516742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49017829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATCTGTCAGAACACTGCCATGA[C/T]AAAATTAAAAATCCCTGTGATAATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VSTM4 PubMed Links

Gene Details

Gene
VSTM4
Gene Name
V-set and transmembrane domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031746.4 3251 UTR 3 NP_001026916.2
NM_144984.3 3251 Intron NP_659421.1
XM_017015825.1 3251 Intron XP_016871314.1
XM_017015826.1 3251 Intron XP_016871315.1
XM_017015827.1 3251 Intron XP_016871316.1
XM_017015828.1 3251 UTR 3 XP_016871317.1

View Full Product Details