Product Details

SNP ID
rs144764735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:131923022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAACAGCTTTTGAGCTGTAATTCA[C/T]TGAGCACAGAGCTCATCCTTTGTGA
Phenotype
MIM: 613992
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP2R2D PubMed Links

Gene Details

Gene
PPP2R2D
Gene Name
protein phosphatase 2 regulatory subunit Bdelta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291310.1 Intron NP_001278239.1
NM_018461.4 Intron NP_060931.2
XM_006717914.3 Intron XP_006717977.1
XM_006717915.2 Intron XP_006717978.1
XM_017016406.1 Intron XP_016871895.1
XM_017016407.1 Intron XP_016871896.1
XM_017016408.1 Intron XP_016871897.1

View Full Product Details