Product Details

SNP ID
rs145068570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100988451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGTCACAGTTGCCTGCGGGCA[C/G]TGAGCCCCTTTGCAGAGTCTTCACA
Phenotype
MIM: 606075 MIM: 610454 MIM: 611848
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C10orf2 PubMed Links

Gene Details

Gene
C10orf2
Gene Name
chromosome 10 open reading frame 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163812.1 415 Missense Mutation CTG,GTG L81V NP_001157284.1
NM_001163813.1 415 Intron NP_001157285.1
NM_001163814.1 415 Intron NP_001157286.1
NM_021830.4 415 Missense Mutation CTG,GTG L81V NP_068602.2
XM_011539975.2 415 Intron XP_011538277.1
XM_017016437.1 415 UTR 5 XP_016871926.1
Gene
LZTS2
Gene Name
leucine zipper tumor suppressor 2
There are no transcripts associated with this gene.

Gene
MRPL43
Gene Name
mitochondrial ribosomal protein L43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308396.1 415 Intron NP_001295325.1
NM_032112.2 415 Intron NP_115488.2
NM_176792.2 415 Intron NP_789762.1
NM_176793.1 415 Intron NP_789763.1
NM_176794.1 415 Intron NP_789764.1
XM_005270231.2 415 Intron XP_005270288.1
XM_006718035.3 415 Intron XP_006718098.1
XM_017016790.1 415 Intron XP_016872279.1
Gene
SEMA4G
Gene Name
semaphorin 4G
There are no transcripts associated with this gene.

View Full Product Details