Product Details

SNP ID
rs145329305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93074520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACCTGCACGACTCCTCGCACAA[A/G]CAGCGCAAGAAGGTGGGGGCAGGAG
Phenotype
MIM: 602239 MIM: 608428
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP26A1 PubMed Links

Gene Details

Gene
CYP26A1
Gene Name
cytochrome P450 family 26 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000783.3 447 Silent Mutation AAA,AAG K134K NP_000774.2
NM_057157.2 447 Silent Mutation AAA,AAG K65K NP_476498.1
Gene
CYP26C1
Gene Name
cytochrome P450 family 26 subfamily C member 1
There are no transcripts associated with this gene.

View Full Product Details