Product Details

SNP ID
rs145472895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97436486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGTTGTACTCGGGCTACTTTCC[A/G]GAATTCTTTAGTGTGGGTCTTAGGG
Phenotype
MIM: 606493 MIM: 172250 MIM: 616750
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOSC1 PubMed Links

Gene Details

Gene
EXOSC1
Gene Name
exosome component 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318362.1 487 Missense Mutation CGG,TGG R158W NP_001305291.1
NM_001318363.1 487 Missense Mutation CGG,TGG R140W NP_001305292.1
NM_001318364.1 487 Missense Mutation CGG,TGG R117W NP_001305293.1
NM_001318365.1 487 Missense Mutation CGG,TGG R97W NP_001305294.1
NM_001318366.1 487 Missense Mutation CGG,TGG R97W NP_001305295.1
NM_016046.4 487 Missense Mutation CGG,TGG R183W NP_057130.1
XM_011539847.2 487 Missense Mutation CGG,TGG R148W XP_011538149.1
XM_017016304.1 487 Missense Mutation CGG,TGG R114W XP_016871793.1
Gene
PGAM1
Gene Name
phosphoglycerate mutase 1
There are no transcripts associated with this gene.

Gene
ZDHHC16
Gene Name
zinc finger DHHC-type containing 16
There are no transcripts associated with this gene.

View Full Product Details