Product Details

SNP ID
rs145767283
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110001654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTTGGGGTGCTTAAAAAGTCCT[C/T]AATCGTCTTCTTTATTCAGAGATTT
Phenotype
MIM: 601568
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADD3 PubMed Links

Gene Details

Gene
ADD3
Gene Name
adducin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001121.3 Intron NP_001112.2
NM_001320591.1 Intron NP_001307520.1
NM_001320592.1 Intron NP_001307521.1
NM_001320593.1 Intron NP_001307522.1
NM_001320594.1 Intron NP_001307523.1
NM_016824.4 Intron NP_058432.1
NM_019903.4 Intron NP_063968.1
Gene
ADD3-AS1
Gene Name
ADD3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details