Product Details

SNP ID
rs145922303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49819195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGTACCAACTGACAACTGCACA[C/T]GTGTGGAAGAGATTGCGTCCTTGAC
Phenotype
MIM: 603501
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARG PubMed Links

Gene Details

Gene
PARG
Gene Name
poly(ADP-ribose) glycohydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303486.1 3378 UTR 3 NP_001290415.1
NM_001303487.1 3378 UTR 3 NP_001290416.1
NM_001303489.1 3378 UTR 3 NP_001290418.1
NM_001324381.1 3378 UTR 3 NP_001311310.1
NM_003631.3 3378 UTR 3 NP_003622.2
XM_011540305.1 3378 UTR 3 XP_011538607.1
XM_011540308.2 3378 Intron XP_011538610.1

View Full Product Details