Product Details

SNP ID
rs145975565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96981867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGATAAATGATTATGATAACCA[A/G]TGTGATGTTGTTTATATCAGTCAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf12 PubMed Links

Gene Details

Gene
C10orf12
Gene Name
chromosome 10 open reading frame 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015652.2 584 Silent Mutation CAA,CAG Q159Q NP_056467.2

View Full Product Details