Product Details

SNP ID
rs145993159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73773237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGAATCGGGAGCTGCCTACCGC[A/G]CGGCTACAGGACACAGCCACGGCCA
Phenotype
MIM: 608842 MIM: 616932 MIM: 607185
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHCHD1 PubMed Links

Gene Details

Gene
CHCHD1
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 1
There are no transcripts associated with this gene.

Gene
FUT11
Gene Name
fucosyltransferase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284194.1 915 Silent Mutation GCA,GCG A252A NP_001271123.1
NM_173540.2 915 Silent Mutation GCA,GCG A252A NP_775811.2
XM_006717656.3 915 Silent Mutation GCA,GCG A252A XP_006717719.1
Gene
SEC24C
Gene Name
SEC24 homolog C, COPII coat complex component
There are no transcripts associated with this gene.

View Full Product Details