Product Details

SNP ID
rs146130725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112153580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCACAAAACTCAGAATATATTC[G/T]AGAAGTTTTTGTCGGTTGCATTGAG
Phenotype
MIM: 602395
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPAM PubMed Links

Gene Details

Gene
GPAM
Gene Name
glycerol-3-phosphate acyltransferase, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244949.1 2667 Silent Mutation CTA,CTC L819L NP_001231878.1
NM_020918.5 2667 Silent Mutation CTA,CTC L819L NP_065969.3
XM_005269998.1 2667 Silent Mutation CTA,CTC L819L XP_005270055.1

View Full Product Details