Product Details

SNP ID
rs146332380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:126426071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCTTCTTTTTTCTTTTTAACTT[A/C]CGGCAAGTTATCATAGATTCTGAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C10orf90 PubMed Links

Gene Details

Gene
C10orf90
Gene Name
chromosome 10 open reading frame 90
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004298.2 2147 Missense Mutation NP_001004298.2
XM_005269491.1 2147 Nonsense Mutation XP_005269548.1
XM_005269492.2 2147 Nonsense Mutation XP_005269549.1
XM_011539216.1 2147 Nonsense Mutation XP_011537518.1
XM_011539217.2 2147 Nonsense Mutation XP_011537519.1
XM_011539220.2 2147 Nonsense Mutation XP_011537522.1
XM_011539222.2 2147 Nonsense Mutation XP_011537524.1
XM_011539223.2 2147 Nonsense Mutation XP_011537525.1
XM_011539224.2 2147 Nonsense Mutation XP_011537526.1
XM_011539226.1 2147 Nonsense Mutation XP_011537528.1
XM_011539227.2 2147 Nonsense Mutation XP_011537529.1
XM_011539230.2 2147 Missense Mutation XP_011537532.1
XM_011539231.1 2147 Nonsense Mutation XP_011537533.1
XM_011539232.1 2147 Nonsense Mutation XP_011537534.1
XM_011539233.1 2147 Nonsense Mutation XP_011537535.1
XM_017015634.1 2147 Nonsense Mutation XP_016871123.1
XM_017015635.1 2147 Intron XP_016871124.1
Gene
LINC00601
Gene Name
long intergenic non-protein coding RNA 601
There are no transcripts associated with this gene.

Gene
LOC728158
Gene Name
hCG2044975
There are no transcripts associated with this gene.

View Full Product Details