Product Details

SNP ID
rs146360818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96981832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGACAGCTCGGAAAAGTAAAAGG[C/G]CATCAGGGCTGAGGATAAATGATTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C10orf12 PubMed Links

Gene Details

Gene
C10orf12
Gene Name
chromosome 10 open reading frame 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015652.2 549 Missense Mutation CCA,GCA P148A NP_056467.2

View Full Product Details