Product Details

SNP ID
rs146410251
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123157791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGAATACTGTCCAGAAGTGAAT[A/G]TGATGGTCACTGGAAGTTGGGATCA
Phenotype
MIM: 603719 MIM: 600647
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BUB3 PubMed Links

Gene Details

Gene
BUB3
Gene Name
BUB3, mitotic checkpoint protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007793.2 570 Missense Mutation ATG,GTG M110V NP_001007794.1
NM_004725.3 570 Missense Mutation ATG,GTG M110V NP_004716.1
Gene
HMX2
Gene Name
H6 family homeobox 2
There are no transcripts associated with this gene.

View Full Product Details