Product Details

SNP ID
rs146428421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68099226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATTAGACTAAATTAATAATGT[C/T]AACTTGAAAATCTGAGTTTTACTGC
Phenotype
MIM: 608517
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYPN PubMed Links

Gene Details

Gene
MYPN
Gene Name
myopalladin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256267.1 Intron NP_001243196.1
NM_001256268.1 Intron NP_001243197.1
NM_032578.3 Intron NP_115967.2
XM_017016833.1 Intron XP_016872322.1
XM_017016834.1 Intron XP_016872323.1

View Full Product Details