Product Details
- SNP ID
-
rs146495217
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:124447670 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGGCTGGGGCGCGCCACCCCCC[A/G]CCTGCGCCGCGCCGTCGGCACCCGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NKX1-2
PubMed Links
Gene Details
- Gene
- NKX1-2
- Gene Name
- NK1 homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001146340.1 |
730 |
Missense Mutation |
GCG,GTG |
A231V |
NP_001139812.1 |
View Full Product Details