Product Details

SNP ID
rs146562360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92834785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACCTATGAGAATGTCCTGGCTG[A/G]GATTCAGTCTTTCGAACTGCCTGTT
Phenotype
MIM: 609672
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOC6 PubMed Links

Gene Details

Gene
EXOC6
Gene Name
exocyst complex component 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013848.3 248 Missense Mutation GAG,GGG E16G NP_001013870.1
NM_001319194.1 248 Missense Mutation GAG,GGG E37G NP_001306123.1
NM_001319195.1 248 Intron NP_001306124.1
NM_001319200.1 248 Intron NP_001306129.1
NM_019053.5 248 Intron NP_061926.3
XM_017016344.1 248 Missense Mutation GAG,GGG E37G XP_016871833.1
XM_017016345.1 248 Missense Mutation GAG,GGG E16G XP_016871834.1
XM_017016346.1 248 Missense Mutation GAG,GGG E16G XP_016871835.1
XM_017016347.1 248 Intron XP_016871836.1
XM_017016348.1 248 Missense Mutation GAG,GGG E37G XP_016871837.1

View Full Product Details