Product Details

SNP ID
rs146576556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112284650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGCTGTGTTACAATGGGGTCCA[C/T]GAAGGAGGTTACTACCAATTTGTGA
Phenotype
MIM: 602653
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TECTB PubMed Links

Gene Details

Gene
TECTB
Gene Name
tectorin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058222.2 265 Silent Mutation CAC,CAT H64H NP_478129.1
XM_017016604.1 265 Silent Mutation CAC,CAT H64H XP_016872093.1

View Full Product Details