Product Details

SNP ID
rs146768172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43375502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTCTCTGTACCCACCCCAGACT[A/G]GAAAAACCTGCAGCTGAGCGGACTG
Phenotype
MIM: 616926
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FXYD4 PubMed Links

Gene Details

Gene
FXYD4
Gene Name
FXYD domain containing ion transport regulator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184963.1 433 Nonsense Mutation TAG,TGG *34W NP_001171892.1
NM_173160.2 433 Nonsense Mutation TAG,TGG *34W NP_775183.1

View Full Product Details