Product Details
- SNP ID
-
hCV160705209
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:47367895 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGCAGGCAAGGGCCTCTTCTCTC[T/C]GCTTCTGAGAATGTGGGTCAGGCCC
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF488
PubMed Links
Gene Details
- Gene
- ZNF488
- Gene Name
- zinc finger protein 488
There are no transcripts associated with this gene.
View Full Product Details