Product Details

SNP ID
rs146852103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15212739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCGCCAAAGTCATCCTTTATTCC[A/G]AGTAATAACTTTAATTCCTTTCTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM171A1 PubMed Links

Gene Details

Gene
FAM171A1
Gene Name
family with sequence similarity 171 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010924.1 2777 UTR 3 NP_001010924.1
XM_011519378.2 2777 UTR 3 XP_011517680.1
XM_017015904.1 2777 UTR 3 XP_016871393.1
XM_017015905.1 2777 UTR 3 XP_016871394.1

View Full Product Details