Product Details

SNP ID
rs146874803
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122850513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGATACCACCAGCGATGACAGG[A/C]ATAATCACTGTATGGAGGTCAAAAG
Phenotype
MIM: 616644
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CUZD1 PubMed Links
Additional Information
For this assay, SNP(s) [rs1891110] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CUZD1
Gene Name
CUB and zona pellucida like domains 1
There are no transcripts associated with this gene.

Gene
FAM24B
Gene Name
family with sequence similarity 24 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204364.1 307 Missense Mutation ATG,ATT M1I NP_001191293.1
NM_152644.2 307 Missense Mutation ATG,ATT M1I NP_689857.2
Gene
FAM24B-CUZD1
Gene Name
FAM24B-CUZD1 readthrough
There are no transcripts associated with this gene.

View Full Product Details