Product Details

SNP ID
rs147163319
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12898042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTGGGGAGCTGGGAACCACCCT[A/G]ATTTTTTTCTGAAATAAAGGCTAAG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CCDC3 PubMed Links

Gene Details

Gene
CCDC3
Gene Name
coiled-coil domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282658.1 1321 UTR 3 NP_001269587.1
NM_031455.3 1321 UTR 3 NP_113643.1

View Full Product Details