Product Details

SNP ID
rs147182520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68901439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGAGCTGGAAGCACCCTTGGAG[A/G]AGTCCGAGAGCCAGAAGAAGGAGAG
Phenotype
MIM: 610373 MIM: 609397
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX50 PubMed Links

Gene Details

Gene
DDX50
Gene Name
DEAD-box helicase 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024045.1 160 Missense Mutation AAG,GAG K19E NP_076950.1
XM_005270148.2 160 UTR 5 XP_005270205.1
XM_011540144.2 160 UTR 5 XP_011538446.1
XM_017016626.1 160 Missense Mutation AAG,GAG K19E XP_016872115.1
XM_017016627.1 160 Intron XP_016872116.1
XM_017016628.1 160 Intron XP_016872117.1
XM_017016629.1 160 Intron XP_016872118.1
XM_017016630.1 160 UTR 5 XP_016872119.1
Gene
STOX1
Gene Name
storkhead box 1
There are no transcripts associated with this gene.

View Full Product Details