Product Details

SNP ID
rs147251818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70433021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATCCACATACAGCATGCTCAGC[A/G]GCTTGGTCTTCACTGGGGCACAACA
Phenotype
MIM: 602224 MIM: 601265
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4EBP2 PubMed Links

Gene Details

Gene
EIF4EBP2
Gene Name
eukaryotic translation initiation factor 4E binding protein 2
There are no transcripts associated with this gene.

Gene
NODAL
Gene Name
nodal growth differentiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018055.4 1001 Missense Mutation CCG,CTG P320L NP_060525.3

View Full Product Details