Product Details

SNP ID
rs147366412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:25175667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGAGGAGGTGCCCATGGACGTG[A/G]CCTCTTACCTCTACACCGGGGACTC
Phenotype
MIM: 614573
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR158 PubMed Links

Gene Details

Gene
GPR158
Gene Name
G protein-coupled receptor 158
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020752.2 307 Missense Mutation ACC,GCC T83A NP_065803.2
XM_017016452.1 307 Intron XP_016871941.1
Gene
GPR158-AS1
Gene Name
GPR158 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details