Product Details

SNP ID
rs147443532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122276794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCTTTGATTACCAAGCAGCAC[A/G]AAGCTCGGCTGGAACGCCGGGTCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTBD16 PubMed Links

Gene Details

Gene
BTBD16
Gene Name
BTB domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318189.1 283 Missense Mutation AAA,GAA K9E NP_001305118.1
NM_144587.3 283 Missense Mutation AAA,GAA K8E NP_653188.2
XM_011539239.2 283 Missense Mutation AAA,GAA K9E XP_011537541.1
XM_011539240.2 283 Missense Mutation AAA,GAA K9E XP_011537542.1
XM_011539241.2 283 Missense Mutation AAA,GAA K9E XP_011537543.1
XM_011539242.2 283 Missense Mutation AAA,GAA K9E XP_011537544.1
XM_017015636.1 283 Missense Mutation AAA,GAA K9E XP_016871125.1
XM_017015637.1 283 Intron XP_016871126.1

View Full Product Details